siRNA Details
| Gene Symbol: Cd177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026862.3 | NP_081138.2 | 11 | 1367 | ||
| GenBank mRNA | BC005615.1 | - | 6 | 737 | ||
| BC027283.1 | - | 11 | 1367 | 2648499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd177| Gene Symbol: Cd177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026862.3 | NP_081138.2 | 11 | 1367 | ||
| GenBank mRNA | BC005615.1 | - | 6 | 737 | ||
| BC027283.1 | - | 11 | 1367 | 2648499 | ||