siRNA Details
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018346.1 | - | 7 | 622 | ||
| AK041864.1 | - | 7 | 618 | |||
| AK082629.1 | - | 7 | 625 | |||
| AK158546.1 | - | 7 | 615 | |||
| AK169761.1 | - | 7 | 622 | |||
| BC016073.1 | - | 7 | 566 | 4482130 | ||

Cdkal1
Genomic Map