siRNA Details
| Gene Symbol: Aspscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164224.1 | NP_001157696.1 | 2 | 79 | ||
| NM_026877.2 | NP_081153.1 | 3 | 300 | |||
| NM_198223.2 | NP_937866.1 | 2 | 79 | |||
| GenBank mRNA | AK088197.1 | - | 3 | 151 | ||
| AK146061.1 | - | 2 | 80 | |||
| AY349132.1 | - | 1 | 69 | |||
| AY349133.1 | - | 3 | 217 | |||
| BC019177.1 | - | 3 | 243 | 5008127 | ||
| BC022115.1 | - | 3 | 247 | 5101238 | ||
| BC031153.1 | - | 3 | 232 | 4976569 | ||

Aspscr1
Genomic Map