siRNA Details
| Gene Symbol: Chmp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026879.2 | NP_081155.1 | 3 | 356 | ||
| GenBank mRNA | AK084111.1 | - | 3 | 358 | ||
| AK180291.1 | - | 3 | 175 | |||
| AK197280.1 | - | 3 | 175 | |||
| AK201812.1 | - | 3 | 173 | |||
| AK217905.1 | - | 3 | 176 | |||
| BC042626.1 | - | 2 | 112 | |||
| BC055809.1 | - | 3 | 385 | 6414077 | ||

Chmp2b
Genomic Map