siRNA Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026883.3 | NP_081159.1 | 1, 2 | 84 | ||
| GenBank mRNA | AK005211.1 | - | 1, 2 | 476 | ||
| AK150218.1 | - | 1, 2 | 483 | |||
| AK193505.1 | - | 12 | ||||
| BC019498.1 | - | 1, 2 | 442 | 4460879 | ||
| BC057658.1 | - | 1, 2 | 123 | 3153717 | ||
| BC061033.1 | - | 1, 2 | 84 | 6590939 | ||

Fam216a
Genomic Map