siRNA Details
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026893.3 | NP_081169.2 | 8 | 1465 | ||
GenBank mRNA | AK005099.1 | - | 8 | 1411 | ||
AK030173.1 | - | 8 | 1417 | |||
AK031089.1 | - | 8 | 1395 | |||
AK087517.1 | - | 8 | 1349 | |||
AK143635.1 | - | 8 | 1419 | |||
AK173289.1 | - | 8 | 1325 | |||
BC023852.1 | - | 8 | 1401 | 5346910 |
