siRNA Details
| Gene Symbol: Dcaf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026893.3 | NP_081169.2 | 3 | 763 | ||
| GenBank mRNA | AK005099.1 | - | 3 | 709 | ||
| AK030173.1 | - | 3 | 715 | |||
| AK031089.1 | - | 3 | 693 | |||
| AK078036.1 | - | 1 | 64 | |||
| AK087517.1 | - | 3 | 647 | |||
| AK090028.1 | - | 3 | 712 | |||
| AK143635.1 | - | 3 | 716 | |||
| AK173289.1 | - | 3 | 623 | |||
| AK188223.1 | - | 2 | 271 | |||
| BC023852.1 | - | 3 | 699 | 5346910 | ||

Dcaf12
Genomic Map