siRNA Details
| Gene Symbol: Snrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021336.4 | NP_067311.4 | 6 | 783 | ||
| GenBank mRNA | AK088438.1 | - | 6 | 558 | ||
| AK151800.1 | - | 6 | 576 | |||
| AK152546.1 | - | 6 | 576 | |||
| AK153502.1 | - | 6 | 576 | |||
| AK206837.1 | - | 5 | 346 | |||
| BC013777.1 | - | 6 | 550 | 2651704 | ||
| BC029642.1 | - | 6 | 550 | 3588138 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271323, XM_006229323, FQ232647.

Snrpa1
Genomic Map