siRNA Details
Gene Symbol: Zg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026918.2 | NP_081194.1 | 3 | 459 | ||
GenBank mRNA | AK007426.1 | - | 1 | 50 | ||
BC031800.1 | - | 3 | 459 | 4208847 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026918.2 | NP_081194.1 | 3 | 459 | ||
GenBank mRNA | AK007426.1 | - | 1 | 50 | ||
BC031800.1 | - | 3 | 459 | 4208847 |