siRNA Details
| Gene Symbol: Kdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083916.1 | NP_001077385.1 | 2 | 897 | ||
| NM_133707.2 | NP_598468.2 | 2 | 838 | |||
| GenBank mRNA | AK007551.1 | - | 1 | 763 | ||
| AK158383.1 | - | 2 | 984 | |||
| BC006890.1 | - | 2 | 766 | 3599314 | ||

Kdf1
Genomic Map