siRNA Details
Gene Symbol: A730017L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042707.1 | - | 4 | 850 |
Gene Symbol: Psmd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178616.3 | NP_848731.2 | 10, 11 | 1087 | ||
GenBank mRNA | AK007547.1 | - | 10, 11 | 1056 | ||
AK029365.1 | - | 6, 7 | 658 | |||
AK051738.1 | - | 10, 11 | 1077 | |||
AK076002.1 | - | 10, 11 | 1075 | |||
AK089942.1 | - | 10, 11 | 1052 | |||
AK136049.1 | - | 10, 11 | 1050 | |||
AK140773.1 | - | 1072 | ||||
AK167559.1 | - | 10, 11 | 1058 | |||
AK168289.1 | - | 10, 11 | 1079 | |||
BC030432.1 | - | 7, 8 | 741 | |||
BC037590.1 | - | 10, 11 | 1036 | |||
BC055457.1 | - | 10, 11 | 1019 | |||
BC090664.1 | - | 10, 11 | 1039 | 30677672 | ||
BC090980.1 | - | 10, 11 | 1054 | 30283128 | ||
BC100595.1 | - | 10, 11 | 1049 | 6767671 | ||
BC119136.1 | - | 10, 11 | 1058 | 8733885 | ||
BC119138.1 | - | 10, 11 | 1058 | 8733887 |
