siRNA Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199187.2 | NP_001186116.1 | 4 | 397 | ||
| NM_026937.3 | NP_081213.1 | 4 | 410 | |||
| GenBank mRNA | AK007519.1 | - | 4 | 394 | ||
| AK053811.1 | - | 4 | 411 | |||
| AK139944.1 | - | 4 | 397 | |||
| AK144614.1 | - | 4 | 371 | |||
| AK171958.1 | - | 4 | 371 | |||
| AK201964.1 | - | 3 | 254 | |||
| BC030905.1 | - | 4 | 376 | 4503445 | ||

Ascc1
Genomic Map