siRNA Details
| Gene Symbol: Chrdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291320.1 | NP_001278249.1 | 7 | 834 | ||
| NM_133709.3 | NP_598470.3 | 7 | 876 | |||
| GenBank mRNA | AF338222.1 | - | 7 | 780 | ||
| AK007577.1 | - | 7 | 839 | |||
| BC019399.1 | - | 7 | 841 | 4012143 | ||

Chrdl2
Genomic Map