siRNA Details
| Gene Symbol: Chrdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133709.3 | NP_598470.3 | 1 | 198 | ||
| GenBank mRNA | AF338222.1 | - | 1 | 102 | ||
| BC019399.1 | - | 1 | 163 | 4012143 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC127066.1 | - | 405 | Chr. - - [-] on Build | ||

Chrdl2
Genomic Map