siRNA Details
| Gene Symbol: Cdk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109626.1 | NP_001103096.1 | 13 | 3453 | ||
| NM_001109628.1 | NP_001103098.1 | 13 | 3453 | |||
| NM_026952.2 | NP_081228.2 | 13 | 3453 | |||
| GenBank mRNA | AK049376.1 | - | 13 | 3100 | ||
| AK129237.1 | - | 13 | 2592 | |||
| AK137734.1 | - | 11 | 1323 | |||
| AY072295.1 | - | 13 | 3444 | |||
| BC057057.1 | - | 3 | 400 | |||
| BC116645.1 | - | 13 | 3436 | 40086381 | ||

Cdk12
Genomic Map