siRNA Details
Gene Symbol: Snx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199188.1 | NP_001186117.1 | 6, 7 | 742 | ||
NM_024225.5 | NP_077187.1 | 5, 6 | 653 | |||
GenBank mRNA | AK003091.1 | - | 366 | |||
AK007676.1 | - | 5, 6 | 601 | |||
AK029463.1 | - | 5, 6 | 613 | |||
AK031873.1 | - | 6, 7 | 634 | |||
AK049129.1 | - | 6, 7 | 634 | |||
AK078776.1 | - | 6, 7 | 635 | |||
AK144710.1 | - | 6, 7 | 635 | |||
AK152311.1 | - | 6, 7 | 678 | |||
AK159115.1 | - | 2, 3 | 196 | |||
AK159904.1 | - | 5, 6 | 558 | |||
AK167362.1 | - | 5, 6 | 573 | |||
AK169786.1 | - | 6, 7 | 634 | |||
AK203820.1 | - | 1, 2 | 31 | |||
BC002242.1 | - | 6, 7 | 711 | 3492262 |
