siRNA Details
| Gene Symbol: Dyrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014390.2 | NP_001014412.1 | 3 | 723 | ||
| GenBank mRNA | AK149371.1 | - | 3 | 959 | ||
| AK155496.1 | - | 3 | 972 | |||
| BC085145.1 | - | 3 | 723 | 6808145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dyrk2| Gene Symbol: Dyrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014390.2 | NP_001014412.1 | 3 | 723 | ||
| GenBank mRNA | AK149371.1 | - | 3 | 959 | ||
| AK155496.1 | - | 3 | 972 | |||
| BC085145.1 | - | 3 | 723 | 6808145 | ||