siRNA Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027727.2 | NP_082003.1 | 2, 3 | 408 | ||
| GenBank mRNA | AK016957.1 | - | 2, 3 | 337 | ||
| AK049909.1 | - | 2, 3 | 373 | |||
| AK080149.1 | - | 2, 3 | 385 | |||
| AK081242.1 | - | 2, 3 | 400 | |||
| AK149628.1 | - | 2, 3 | 382 | |||
| AK149879.1 | - | 2, 3 | 383 | |||
| AK149891.1 | - | 2, 3 | 382 | |||
| AK154794.1 | - | 2, 3 | 397 | |||
| AK199364.1 | - | 19 | ||||
| BC038276.1 | - | 2, 3 | 410 | 4458792 | ||
| BC092283.1 | - | 2, 3 | 394 | |||

Dym
Genomic Map