siRNA Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027727.2 | NP_082003.1 | 15, 16 | 2011 | ||
| GenBank mRNA | AK007748.1 | - | 3, 4 | 580 | ||
| AK049909.1 | - | 15, 16 | 1976 | |||
| AK154794.1 | - | 15, 16 | 2000 | |||
| AK198427.1 | - | 3, 4 | 325 | |||
| BC018220.1 | - | 11, 12 | 1371 | 3985017 | ||
| BC038276.1 | - | 15, 16 | 2013 | 4458792 | ||

Dym
Genomic Map