siRNA Details
| Gene Symbol: Sat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026991.2 | NP_081267.1 | 1 | 197 | ||
| GenBank mRNA | AK011406.1 | - | 2 | 115 | ||
| AK161230.1 | - | 1 | 255 | |||
| BC061227.1 | - | 1 | 197 | 30312881 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sat2| Gene Symbol: Sat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026991.2 | NP_081267.1 | 1 | 197 | ||
| GenBank mRNA | AK011406.1 | - | 2 | 115 | ||
| AK161230.1 | - | 1 | 255 | |||
| BC061227.1 | - | 1 | 197 | 30312881 | ||