siRNA Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040396.2 | NP_001035486.2 | 3 | 340 | ||
| GenBank mRNA | AK013022.1 | - | 3 | 304 | ||
| AK032838.1 | - | 3 | 317 | |||
| AK047958.1 | - | 3 | 318 | |||
| AK146778.1 | - | 3 | 228 | |||
| BC019970.1 | - | 3 | 288 | 4458171 | ||
| BC038867.1 | - | 3 | 216 | 1547418 | ||
| BC043068.1 | - | 226 | ||||
| BC132453.1 | - | 3 | 306 | 40130730 | ||
| BC138424.1 | - | 3 | 306 | 8861444 | ||
| CJ133950.1 | - | 3 | 341 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001014253, BC086953, AY995234.

Selt
Genomic Map