siRNA Details
| Gene Symbol: Zfp688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026999.4 | NP_081275.3 | 3 | 599 | ||
| GenBank mRNA | AK013037.1 | - | 2 | 879 | ||
| AK170857.1 | - | 2 | 2321 | |||
| BC027683.1 | - | 2 | 882 | 1345974 | ||
| BC038680.1 | - | 1 | 170 | |||
| BC052512.1 | - | 3 | 439 | 1362502 | ||
| BQ550868.1 | - | 3 | 508 | |||

Zfp688
Genomic Map