siRNA Details
| Gene Symbol: Slc16a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172371.3 | NP_758959.1 | 1 | 425 | ||
| GenBank mRNA | AK028143.1 | - | 1 | 410 | ||
| AK028759.1 | - | 1 | 426 | |||
| BC039949.1 | - | 1 | 213 | 4167719 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a13| Gene Symbol: Slc16a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172371.3 | NP_758959.1 | 1 | 425 | ||
| GenBank mRNA | AK028143.1 | - | 1 | 410 | ||
| AK028759.1 | - | 1 | 426 | |||
| BC039949.1 | - | 1 | 213 | 4167719 | ||