siRNA Details
Gene Symbol: Pacrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027032.2 | NP_081308.1 | 5 | 1001 | ||
GenBank mRNA | AK005771.1 | - | 5 | 997 | ||
AK135510.1 | - | 5 | 1003 | |||
BC120740.1 | - | 5 | 979 | 40129663 |
