siRNA Details
| Gene Symbol: Ppp1r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145692.2 | NP_663730.1 | 4 | 531 | ||
| GenBank mRNA | AK005871.1 | - | 4 | 526 | ||
| AK015164.1 | - | 4 | 526 | |||
| AK076637.1 | - | 4 | 532 | |||
| BC022722.1 | - | 4 | 515 | 4503172 | ||

Ppp1r42
Genomic Map