siRNA Details
| Gene Symbol: Slc38a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027052.3 | NP_081328.2 | 3 | 271 | ||
| GenBank mRNA | AB055004.1 | - | 3 | 247 | ||
| AK051023.1 | - | 3 | 272 | |||
| AK164292.1 | - | 3 | 214 | |||
| AK167539.1 | - | 3 | 253 | |||
| AK169349.1 | - | 2 | 260 | |||
| AY027919.1 | - | 3 | 310 | |||
| BC024072.1 | - | 3 | 196 | 5097293 | ||
| BC024123.1 | - | 3 | 228 | 5098067 | ||
| BC031717.1 | - | 3 | 189 | 4195569 | ||

Slc38a4
Genomic Map