siRNA Details
| Gene Symbol: Zpbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166494.1 | NP_001159966.1 | 7 | 1223 | ||
| NM_001166495.1 | NP_001159967.1 | 8 | 1012 | |||
| NM_027061.2 | NP_081337.2 | 8 | 1103 | |||
| NM_199419.3 | NP_955451.2 | 7 | 884 | |||
| GenBank mRNA | AK006058.1 | - | 7 | 1171 | ||
| AY251601.1 | - | 8 | 1048 | |||
| AY251602.1 | - | 7 | 829 | |||
| BC050986.1 | - | 8 | 979 | |||
| BC089476.1 | - | 7 | 884 | 6708062 | ||

Zpbp2
Genomic Map