siRNA Details
| Gene Symbol: Dnajb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153527.2 | NP_705755.2 | 7 | 826 | ||
| GenBank mRNA | AF419292.2 | - | 7 | 971 | ||
| AK005984.1 | - | 7 | 843 | |||
| AK139680.1 | - | 799 | ||||
| BC048501.1 | - | 7 | 826 | 6705915 | ||

Dnajb13
Genomic Map