siRNA Details
| Gene Symbol: Commd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178377.4 | NP_848464.1 | 2, 3 | 151 | ||
| GenBank mRNA | AB052770.1 | - | 2, 3 | 164 | ||
| AK139321.1 | - | 167 | ||||
| AK159099.1 | - | 2, 3 | 167 | |||
| AK166907.1 | - | 2, 3 | 165 | |||
| BC034879.1 | - | 2, 3 | 151 | 3468632 | ||

Commd10
Genomic Map