siRNA Details
| Gene Symbol: Cdk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081058.2 | NP_001074527.1 | 12 | 3555 | ||
| NM_027118.1 | NP_081394.1 | 12 | 3555 | |||
| GenBank mRNA | AK035493.1 | - | 12 | 2226 | ||
| AK173265.1 | - | 12 | 3555 | |||
| BC051093.1 | - | 5 | 439 | |||
| BC076619.1 | - | 13 | 2010 | |||

Cdk13
Genomic Map