siRNA Details
| Gene Symbol: Odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027128.2 | NP_081404.2 | 4 | 265 | ||
| GenBank mRNA | EF113909.1 | - | 4 | 265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odam| Gene Symbol: Odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027128.2 | NP_081404.2 | 4 | 265 | ||
| GenBank mRNA | EF113909.1 | - | 4 | 265 | ||