siRNA Details
| Gene Symbol: Ap5s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291031.1 | NP_001277960.1 | 3 | 703 | ||
| NM_001291032.1 | NP_001277961.1 | 3 | 703 | |||
| NM_027129.3 | NP_081405.2 | 3 | 703 | |||
| GenBank mRNA | AK002807.1 | - | 3 | 628 | ||
| AK009634.1 | - | 3 | 640 | |||
| BC048399.1 | - | 3 | 623 | 6438951 | ||
| CX204398.1 | - | 2 | 330 | |||

Ap5s1
Genomic Map