siRNA Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110209.1 | NP_001103679.1 | 14 | 1497 | ||
| NM_027133.3 | NP_081409.1 | 13 | 1414 | |||
| GenBank mRNA | AK014897.1 | - | 14 | 1394 | ||
| AK048501.1 | - | 4 | 1657 | |||
| AK082900.1 | - | 12 | 1339 | |||
| AK136506.1 | - | 13 | 1389 | |||
| AK137445.1 | - | 14 | 1497 | |||
| AK173241.1 | - | 13 | 1346 | |||
| AY312281.1 | - | 13 | 1335 | |||
| BC057961.1 | - | 13 | 1341 | 5353629 | ||
| BC060153.1 | - | 12 | 1253 | |||

Lnp
Genomic Map