siRNA Details
| Gene Symbol: Mtfmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027134.3 | NP_081410.2 | 7 | 855 | ||
| GenBank mRNA | AK009430.1 | - | 7 | 856 | ||
| AK133492.1 | - | 7 | 850 | |||
| BC019509.1 | - | 7 | 845 | 4456981 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtfmt| Gene Symbol: Mtfmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027134.3 | NP_081410.2 | 7 | 855 | ||
| GenBank mRNA | AK009430.1 | - | 7 | 856 | ||
| AK133492.1 | - | 7 | 850 | |||
| BC019509.1 | - | 7 | 845 | 4456981 | ||