siRNA Details
| Gene Symbol: Pitrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145131.1 | NP_660113.1 | 24 | 2679 | ||
| GenBank mRNA | AF513714.1 | - | 24 | 2679 | ||
| AK129292.1 | - | 24 | 2653 | |||
| AK141277.1 | - | 24 | 2674 | |||
| AK147837.1 | - | 24 | 2670 | |||
| AK164235.1 | - | 24 | 2641 | |||
| AK167426.1 | - | 24 | 2670 | |||
| AK168335.1 | - | 23 | 2556 | |||
| AY779273.1 | - | 24 | 2679 | |||
| AY779274.1 | - | 24 | 2679 | |||
| BC006917.1 | - | 14 | 1480 | 3601081 | ||

Pitrm1
Genomic Map