siRNA Details
| Gene Symbol: Fam83g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178618.3 | NP_848733.2 | 4 | 1455 | ||
| GenBank mRNA | AK028566.1 | - | 4 | 1402 | ||
| BC085279.1 | - | 4 | 1416 | 30438273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam83g| Gene Symbol: Fam83g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178618.3 | NP_848733.2 | 4 | 1455 | ||
| GenBank mRNA | AK028566.1 | - | 4 | 1402 | ||
| BC085279.1 | - | 4 | 1416 | 30438273 | ||