siRNA Details
| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027168.2 | NP_081444.1 | 2 | 97 | ||
| GenBank mRNA | AK009957.1 | - | 2 | 38 | ||
| AK161186.1 | - | 1 | 1128 | |||
| BC104360.1 | - | 2 | 97 | 40049707 | ||
| BC104361.1 | - | 2 | 97 | 40049709 | ||

Hddc2
Genomic Map