siRNA Details
| Gene Symbol: Pof1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181579.1 | NP_853557.1 | 14 | 1596 | ||
| GenBank mRNA | AF309775.1 | - | 13 | 1489 | ||
| AK036922.1 | - | 14 | 1603 | |||
| AK137653.1 | - | 14 | 1595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pof1b| Gene Symbol: Pof1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181579.1 | NP_853557.1 | 14 | 1596 | ||
| GenBank mRNA | AF309775.1 | - | 13 | 1489 | ||
| AK036922.1 | - | 14 | 1603 | |||
| AK137653.1 | - | 14 | 1595 | |||