siRNA Details
| Gene Symbol: Tatdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175151.4 | NP_780360.1 | 8, 9 | 547 | ||
| GenBank mRNA | AK042248.1 | - | 8, 9 | 548 | ||
| AK161972.1 | - | 8, 9 | 632 | |||
| BC034895.1 | - | 5, 6 | 334 | |||
| BC061187.1 | - | 8, 9 | 544 | 30277245 | ||

Tatdn1
Genomic Map