siRNA Details
| Gene Symbol: Camsap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163749.1 | NP_001157221.1 | 8, 9 | 1412 | ||
| NM_027171.3 | NP_081447.2 | 8, 9 | 1409 | |||
| GenBank mRNA | AK081728.1 | - | 8, 9 | 1184 | ||
| AK220401.1 | - | 4, 5 | 554 | |||
| BC043081.1 | - | 8, 9 | 1068 | |||
| BC048787.1 | - | 8, 9 | 1057 | 6489361 | ||
| KC426930.1 | - | 10, 11 | 1131 | |||
| KC426931.1 | - | 10, 11 | 1128 | |||
| KC426932.1 | - | 9, 10 | 1098 | |||
| KC426933.1 | - | 9, 10 | 1095 | |||
| KC426934.1 | - | 9, 10 | 1083 | |||
| KC426935.1 | - | 9, 10 | 1080 | |||
| KC426936.1 | - | 8, 9 | 1050 | |||
| KC426937.1 | - | 8, 9 | 1047 | |||
| KC426938.1 | - | 10, 11 | 1131 | |||
| KC426939.1 | - | 10, 11 | 1128 | |||
| KC426940.1 | - | 9, 10 | 1098 | |||
| KC426941.1 | - | 9, 10 | 1095 | |||
| KC426942.1 | - | 9, 10 | 1083 | |||
| KC426943.1 | - | 9, 10 | 1080 | |||
| KC426944.1 | - | 8, 9 | 1050 | |||
| KC426945.1 | - | 8, 9 | 1047 | |||

Camsap3
Genomic Map