siRNA Details
| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027175.3 | NP_081451.3 | 2 | 339 | ||
| GenBank mRNA | AK010328.1 | - | 1 | 169 | ||
| AK050343.1 | - | 2 | 340 | |||
| AK215302.1 | - | 1 | 110 | |||
| BC018422.1 | - | 2 | 169 | 3593650 | ||

Ndufaf1
Genomic Map