siRNA Details
Gene Symbol: Ndufaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027175.3 | NP_081451.3 | 2 | 498 | ||
GenBank mRNA | AK010328.1 | - | 1 | 328 | ||
AK050343.1 | - | 2 | 499 | |||
AK215302.1 | - | 1 | 269 | |||
BC018422.1 | - | 2 | 328 | 3593650 |
