siRNA Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027184.1 | NP_081460.1 | 6 | 1253 | ||
| GenBank mRNA | AK036084.1 | - | 6 | 1021 | ||
| AK036978.1 | - | 6 | 1213 | |||
| AK039460.1 | - | 1 | 197 | |||
| AK172246.1 | - | 6 | 1345 | |||
| BC052463.1 | - | 6 | 1253 | 6834748 | ||

Ipmk
Genomic Map