siRNA Details
| Gene Symbol: Ttl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027192.2 | NP_081468.1 | 5 | 864 | ||
| GenBank mRNA | AK010374.1 | - | 3 | 240 | ||
| BC018513.1 | - | 5 | 867 | 3597859 | ||
| BC024414.1 | - | 5 | 836 | 4973810 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttl| Gene Symbol: Ttl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027192.2 | NP_081468.1 | 5 | 864 | ||
| GenBank mRNA | AK010374.1 | - | 3 | 240 | ||
| BC018513.1 | - | 5 | 867 | 3597859 | ||
| BC024414.1 | - | 5 | 836 | 4973810 | ||