siRNA Details
| Gene Symbol: Fbxo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153195.2 | NP_694875.2 | 7 | 1211 | ||
| GenBank mRNA | AK010499.1 | - | 5 | 489 | ||
| AK147716.1 | - | 511 | ||||
| AK147797.1 | - | 7 | 1163 | |||
| AK147907.1 | - | 7 | 1130 | |||
| AK150190.1 | - | 7 | 986 | |||
| AK150626.1 | - | 7 | 1169 | |||
| AK151025.1 | - | 7 | 1169 | |||
| AK152513.1 | - | 7 | 1169 | |||
| BC032153.1 | - | 7 | 1153 | 5054087 | ||

Fbxo7
Genomic Map