siRNA Details
| Gene Symbol: Smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133716.3 | NP_598477.2 | 9 | 1485 | ||
| GenBank mRNA | AK007666.1 | - | 1 | 86 | ||
| BC031581.1 | - | 9 | 1474 | |||
| BC032217.1 | - | 9 | 1461 | |||
| BC052413.1 | - | 9 | 1485 | 5720177 | ||
| BM944792.1 | - | 1 | 33 | |||

Smap2
Genomic Map