siRNA Details
| Gene Symbol: Trim32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161782.1 | NP_001155254.1 | 2 | 1957 | ||
| NM_053084.2 | NP_444314.2 | 2 | 1960 | |||
| GenBank mRNA | AF347694.1 | - | 2 | 1928 | ||
| AK154028.1 | - | 2 | 1925 | |||
| AK166358.1 | - | 2 | 1912 | |||
| AX775777.1 | - | 2 | 1940 | |||
| BC034104.1 | - | 2 | 1899 | 4502346 | ||

Trim32
Genomic Map