siRNA Details
| Gene Symbol: Mterf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178051.3 | NP_835152.1 | 2 | 481 | ||
| GenBank mRNA | AK033043.1 | - | 2 | 492 | ||
| AK077142.1 | - | 2 | 481 | |||
| AK079098.1 | - | 2 | 473 | |||
| BC064709.1 | - | 2 | 474 | 30314353 | ||

Mterf4
Genomic Map