siRNA Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027238.2 | NP_081514.1 | 11, 12 | 995 | ||
| GenBank mRNA | AK037208.1 | - | 11, 12 | 996 | ||
| AK049108.1 | - | 11, 12 | 1046 | |||
| AK132655.1 | - | 6, 7 | 446 | |||
| BC151021.1 | - | 11, 12 | 957 | 9087934 | ||

Ttc39b
Genomic Map