siRNA Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027238.2 | NP_081514.1 | 19 | 1763 | ||
| GenBank mRNA | AK037208.1 | - | 19 | 1764 | ||
| AK043383.1 | - | 2 | 501 | |||
| AK132655.1 | - | 14 | 1214 | |||
| BC151021.1 | - | 19 | 1725 | 9087934 | ||

Ttc39b
Genomic Map